missing translation for 'onlineSavingsMsg'
Learn More

Hyoscine EP Impurity C, Mikromol™

Product Code. 18021220
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18021220 25 mg 25mg
17949989 100 mg 100mg
30459457 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18021220 Supplier Mikromol Supplier No. MM0546.060025
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hyoscine EP Impurity C
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo(3.3.1.02,4)non-7-yl 2-Phenylprop-2-enoate (Apohyoscine)
CAS 535-26-2
Analyte or Component Names (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo(3.3.1.02,4)non-7-yl 2-Phenylprop-2-enoate (Apohyoscine)
Shipping Condition Room Temperature
Molecular Formula C17 H19 N O3
Certifications/Compliance ISO 17025
Synonym (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl 2-Phenylprop-2-enoate (Apohyoscine), Hyoscine Hydrobromide Imp. C (EP), Hyoscine Imp. C (EP), (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl 2-Phenylprop-2-enoate, Apohyoscine, Hyoscine Hydrobromide Impurity C, Hyoscine Impurity C
InChI Formula InChI=1S/C17H19NO3/c1-10(11-6-4-3-5-7-11)17(19)20-12-8-13-15-16(21-15)14(9-12)18(13)2/h3-7,12-16H,1,8-9H2,2H3/t12-,13-,14+,15-,16+
SMILES CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(=C)c4ccccc4
IUPAC Name (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl 2-Phenylprop-2-enoate
Molecular Weight (g/mol) 285.34
Quantity 25 mg
Formula Weight 285.1365 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.