missing translation for 'onlineSavingsMsg'
Learn More

Hyoscyamine, Mikromol™

Product Code. 18073790
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073790 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18073790 Supplier Mikromol Supplier No. MM1699.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hyoscyamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hyoscyamine
CAS 101-31-5
Analyte or Component Names Hyoscyamine
Shipping Condition Room Temperature
Molecular Formula C17 H23 N O3
Certifications/Compliance ISO 17034
Synonym 1αH,5αH-Tropan-3α-ol, (-)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, [3(S)-endo]-, Tropic acid, 1αH,5αH-tropan-3α-yl ester, (-)- (8CI), (-)-Atropine, (-)-Hyoscyamine, (S)-(-)-Hyoscyamine, (S)-Atropine, 1αH,5αH-Tropan-3α-yl (-)-tropate, Cystospaz, Daturine, Duboisine, Hyoscyamine, L-Hyoscyamin, L-Hyoscyamine, l-Atropine, l-Hyoscyamine, l-Tropine tropate, Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, (αS)-, Hyoscine Hydrobromide Impurity A, Hyoscine Impurity A
InChI Formula InChI=1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16-/m1/s1
SMILES CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@H](CO)c3ccccc3
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2S)-3-hydroxy-2-phenylpropanoate
Molecular Weight (g/mol) 289.37
Quantity 100 mg
Formula Weight 289.1678 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.