missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYPB Antibody (CL9956) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88941AF647
This item is not returnable.
View return policy
Description
HYPB Monoclonal antibody specifically detects HYPB in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| HYPB | |
| Monoclonal | |
| Alexa Fluor 647 | |
| FLJ22472, FLJ23184, FLJ45883, FLJ46217, HIF1, HIF-1EC 2.1.1.43, HIP-1, histone-lysine N-methyltransferase SETD2, hSET2, huntingtin interacting protein 1, Huntingtin yeast partner B, Huntingtin-interacting protein 1, Huntingtin-interacting protein B, HYPBp231HBP, KIAA1732SET domain-containing protein 2, KMT3AHBP231, Lysine N-methyltransferase 3A, SET domain containing 2, SET2FLJ16420 | |
| This antibody was developed using a synthetic peptide derived from Q9BYW2, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL9956 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 29072 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction