Learn More
BUCHI I-180 Controller for Rotavapor R-80
Product Code. 30000645 Shop All Buchi Products
Change view
Click to view available options
For Use With (Equipment):
Rotavapor R-80 I-100
Rotavapor R-80 V-80/180
Rotavapor R-80 V-80/180 House Vacuum
Includes:
Adapter Cable for I-180, I-100 Power Supply
Communication Cable, Holder
Power Pack
Valve Unit, Woulff Bottle, Communication Cable, Tubing
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Includes Unit Size
30000645 Rotavapor R-80 V-80/180 Communication Cable, Holder Each
30000610 Rotavapor R-80 V-80/180 Power Pack Each
30000632 Rotavapor R-80 V-80/180 House Vacuum Valve Unit, Woulff Bottle, Communication Cable, Tubing Each
30000634 Rotavapor R-80 I-100 Adapter Cable for I-180, I-100 Power Supply Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30000645 Supplier BUCHI Supplier No. 11I1801
  • ONLINE EXCLUSIVE PRICE
    1380.00€
    1100.00€ / Pack of 1
    Save 280.00€
    20% Off
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
The I-180 is an easy-to-use interface for conveniently regulating your vacuum automatically. Increase control of the evaporation process by setting the desired vacuum and accessing other modes of operation.
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
TRUSTED_SUSTAINABILITY
Specifications
Type Controller Interface
For Use With (Equipment) Rotavapor R-80 V-80/180
Includes Communication Cable, Holder