Learn More
Ambion™ mirVana™ miRNA Inhibitor
Product Code. 13403089
Change view
Click to view available options
Quantity:
0.25 nmol
1 nmol
20 nmol
250 nmol
5 nmol
Unit Size:
20nmol
250nmol
5nmol
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13403089 0.25 nmol Each
13413089 1 nmol Each
13496696 5 nmol 5nmol
13416706 20 nmol 20nmol
13426706 250 nmol 250nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13403089 Supplier Ambion™ Supplier No. 4464092
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
mirVana miRNA Inhibitors are single-stranded RNA molecules designed to inhibit endogenous microRNAs (miRNAs) and are ideal for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications.
mirVana miRNA Inhibitors are single-stranded RNA molecules designed to inhibit endogenous microRNAs (miRNAs) and are ideal for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications. Based on the latest version of miRbase, mirVana miRNA Inhibitors are available for a wide range of miRNAs, providing researchers with a powerful tool to investigate gene regulation mechanisms.
Key features include:
  • High specificity and potency—designed to closely resemble natural miRNAs, ensuring high specificity and potent gene regulation
  • Enhanced stability—chemical modifications enhance stability, making them suitable for long-term studies
  • Broad range of targets—available for a comprehensive list of miRNAs, allowing for extensive research possibilities
  • Optimized for delivery—compatible with various transfection reagents and delivery systems, ensuring efficient uptake by cells
miRNA inhibitors to increase gene expression
mirVana miRNA Inhibitors are molecules designed to specifically bind to and inhibit the activity of miRNAs. By inhibiting miRNAs, mirVana miRNA Inhibitors prevent them from binding to their target mRNAs, thereby allowing the expression of those genes to proceed. This can be useful in research to study gene function, understand disease mechanisms, and develop treatments that modulate gene expression.
TRUSTED_SUSTAINABILITY
Tekniske data
Content And Storage Store in freezer (-5 to -30°C).
RNAi Type miRNA
Product Type miRNA
Quantity 0.25 nmol
For Research Use Only. Not for use in diagnostic procedures.
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.