Learn More
Ambion™ mirVana™ miRNA Inhibitor, 1 nmol
Product Code. 13413089
Change view
Click to view available options
1 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13413089 1 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13413089 Supplier Ambion™ Supplier No. 4464093
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
mirVana™ miRNA Inhibitor
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C).
Description mirVana™ miRNA Inhibitor, 1nmol, Small, Chemically Modified Single-Stranded RNA Molecules, Designed to Specifically Bind to and Inhibit Endogenous miRNA, Molecules and Enable miRNA Functional Analysis by Down-Regulation of miRNA Activity
RNAi Type miRNA
Product Type miRNA
Quantity 1 nmol