missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ N-(1-Pyrene)Maleimide

Product Code. 11519256
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
11519256 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 11519256

Brand: Molecular Probes™ P28

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-(1-pyrene)maleimide

The thiol-reactive N-(1-pyrene)maleimide can be used to create environment-sensitive bioconjugates with this unique fluorophore.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Product Type Maleimide
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye Pyrene
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.