Learn More
Brady™ i7500 Industrial Label Printer
Product Code. 30279200
Change view
Click to view available options
Software Type:
Brady WorkStation Basic
Brady WorkStation Suite
Plug Type:
EU
UK
Unit Size:
Each
Includes:
Auto Cutter
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Software Type Plug Type unitSize Includes
30279200 Brady WorkStation Basic EU Each -
30279179 Brady WorkStation Suite EU Each -
30279211 Brady WorkStation Basic EU Each Auto Cutter
30279178 Brady WorkStation Suite EU Each Auto Cutter
30279157 Brady WorkStation Basic UK Each -
30279177 Brady WorkStation Suite UK Each -
30279188 Brady WorkStation Basic UK Each Auto Cutter
30279199 Brady WorkStation Suite UK Each Auto Cutter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Regulatory: This product is under eco-contribution tax.
Product Code. 30279200 Supplier Brady™ Supplier No. 177739
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
High-performance 76.2 mm core label printing the way you never thought possible.
  • No need to set print speed, heat, offsets, sensor mode or sensor location - i7500 does it automatically without wasteful trial-and-error print tests
  • No need to calibrate wasted blank labels - i7500 detects the part size and spacing, and prints on the very first label
  • i75-Series ribbons auto-center with the ink always facing the correct way; the i7500 will warn you if the ink type is a mismatch with the installed label material
  • Over 4,000 stock i75-Series label parts and ribbons in 80+ materials operate in full smart-mode functionality
  • Manual 76.2 mm core label rolls and 25.4 mm core ribbons will also function in full manual mode where the operator makes necessary adjustments for print settings, sensor adjustments, label calibration and correct ribbon pairing
  • Brady WorkStation software (included) auto-detects the installed part for driverless 1-click setup and label file creation; or use Windows driver if using non-Brady software
  • 300 dpi print resolution for day-to-day printing, or user can quickly field-exchange with optional 600 dpi printhead with no calibration or firmware update required
  • Standard configuration ships with a Tear Bar; optional Auto Cutter accessories can be easily user-installed with four fastening screws
Specifications
Product Type Printer
Color Blue, Gray
Display Type Color LCD Touchscreen
Material Plastic
Print Technology Direct Thermal, Thermal Transfer
Length (English) 20.9 in.
Width (English) 11.5 in.
Length (Metric) 530.86 mm
Width (Metric) 292.1 mm
Height (English) 14.6 in.
Height (Metric) 370.84 mm
Dimensions (L x W x H) 292.1 x 370.84 x 530.86 mm
Color Capability Monocolour
Memory 512 MB RAM, 195 MB IFFS Flash
Model i7500-300-EU
Print Speed 304.8 mm/sec.
Resolution 300 dpi
Software Type Brady WorkStation Basic
Connector Types USB, Ethernet, Wi-Fi, Bluetooth
Weight (English) 40 lb.
Weight (Metric) 18.14 kg
Quantity 1 Ea.
Warranty 1 Year
Voltage 100/240 V
Keyboard Type None
Alternate Part Number i7500-300-EU
Plug Type EU