missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ IA

Product Code. 30000430
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000430 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000430 Supplier Tocris Bioscience™ Supplier No. 7789/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SHAPE-JuMP monoadduct control for TBIA (Cat. No. 7788)

IA is a SHAPE-JuMP monoadduct control for TBIA (Cat. No. 7788).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dihydro-3,1-benzoxazine-2,4-dione
CAS 118-48-9
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C8H5NO3
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.