missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IBA1 Polyclonal Antibody, Biotin
GREENER_CHOICE

Product Code. 17886682
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17886682 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17886682 Supplier Invitrogen™ Supplier No. PA5142001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform 1 (NP_116573.1) and isoform 3 (NP_001614.3). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

Ionized calcium-binding adapter molecule 1 (IBA1), also known by its gene name AIF1, is a protein expressed predominantly by microglia in the brain and spinal cord. This protein belongs to the EF-hand calcium-binding protein family and plays a crucial role in microglial activation and migration in response to brain injury or neuroinflammation. IBA1's function is integral to microglial motility and phagocytic activity, facilitating the cellular response to pathogenic stimuli and promoting tissue homeostasis and repair in the central nervous system. IBA1 serves as a reliable marker for activated microglia in various neurological disorders, including Alzheimer's disease, Parkinson's disease, and multiple sclerosis, where increased expression correlates with disease progression and severity. The protein's structural features enable it to bind calcium ions, inducing conformational changes that activate signaling pathways essential for microglial function. Its expression is highly regulated by inflammatory cytokines, underpinning its role in neuroimmune responses. Due to its specific expression in microglia during pathological conditions, IBA1 is widely used in research as a marker to study microglial status and activity, and it remains a focal point for understanding microglial involvement in neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IBA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene AIF1
Gene Accession No. P55008, P55009
Gene Alias AI607846; AIF1; AIF-1; Allograft inflammatory factor 1; balloon angioplasty responsive transcript; balloon angioplasty-responsive transcript 1; Bart1; BART-1; D17H6S50E; DADB-70P7.8; DASS-82G15.2; Em:AF129756.17; G1; Iba; Iba1; interferon gamma responsive transcript; ionized calcium binding adapter molecule 1; ionized calcium-binding adapter molecule 1; IRT1; IRT-1; microglia response factor; Mrf1; MRF-1; protein BART-1; protein G1; testis specific
Gene Symbols AIF1
Host Species Goat
Immunogen Peptide with sequence C-TGPPAKKAISELP.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199, 29427
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.