Learn More
ICA1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560005
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPSASLTSQE PSMGSEPLAH SSRFLPSQLF DLGFHVAGAF NNWVSQEESE LCLSHTDNQP VPSQSPKKLT RSPNNGNQD Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 63%.
ICA1L is a protein coding gene and gene ontology (GO) annotation includes acrosomal vesicle; protein domain specific binding; spermatid development.Specifications
ICA1L | |
Polyclonal | |
Unconjugated | |
ICA1L | |
1700030B17Rik; ALS2CR14; Als2cr15; amyotr; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 14; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 15; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 14 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 15 protein homolog; ICA1L; Ica69-related protein; islet cell autoantigen 1 like; islet cell autoantigen 1, 69kDa-like; islet cell autoantigen 1,69kDa-like; islet cell autoantigen 1-like; islet cell autoantigen 1-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
130026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NDH6 | |
ICA1L | |
Recombinant protein corresponding to Human ICA1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only