missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICA1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15622-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| ICA1L | |
| Polyclonal | |
| Unconjugated | |
| ICA1L | |
| ALS2CR14, ALS2CR15, ICA1L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 130026, 316432, 70375 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TY65, Q6RUG5, Q8NDH6 | |
| Ica1l | |
| ICA1L Fusion Protein Ag8048 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction