Learn More
ICAM-1/CD54 Antibody (MEM-111) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand:  Novus Biologicals NB500-318-25ug
 View more versions of this product
Product Code. 18501231
  • 195.00€ / 25µg
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human, Rat, Bovine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ICAM-1/CD54
Monoclonal
1 mg/mL
Western Blot 1:1000, Flow Cytometry 0.5-4 μg/mL, ELISA 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 10 μg/mL, Immunohistochemistry-Frozen, CyTOF-ready
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Mouse
110 kDa
25 μg
Adaptive Immunity, Cancer, Cell Biology, Immunology
3383
Human, Rat, Bovine
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
MEM-111
Unconjugated
P05362
ICAM1
Raji human Burkitt's lymphoma cell line (NM_000201.2).
Protein A purified
RUO
Primary
The antibody MEM-111 reacts with an extracellular epitope CD54 (ICAM-1), a 85-110 kDa type I transmembrane glycoprotein (receptor for rhinovirus). The expression of CD54 is upregulated by activation; it is expressed on activated endothelial cells, T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells. HLDA VI; WS Code AS A049
Store at 4°C. Do not freeze.
IgG2a