missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1/CD54 Rabbit anti-Mouse, Clone: 9, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00€ - 552.00€
Specifications
| Antigen | ICAM-1/CD54 |
|---|---|
| Clone | 9 |
| Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244078
|
Novus Biologicals
NBP2-90585-50UL |
50 μg |
367.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18259427
|
Novus Biologicals
NBP2-90585-100UL |
100 μg |
552.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| ICAM-1/CD54 | |
| Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| Recombinant Mouse ICAM-1/CD54 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 9 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 3383 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title