missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-3/CD50 Antibody (ICAM3/1019), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47903MFV450
This item is not returnable.
View return policy
Description
ICAM-3/CD50 Monoclonal antibody specifically detects ICAM-3/CD50 in Human samples. It is validated for Flow Cytometry
Specifications
| ICAM-3/CD50 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| ICAM3/1019 | |
| Flow Cytometry | |
| CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
| Recombinant human ICAM-3/CD50 protein (Uniprot: P32942) | |
| 0.1 mL | |
| Cellular Markers, Immunology | |
| 3385 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction