missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ICAT Monoclonal Antibody (536017)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561244

Product Code. 30534250

  • 409.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene binds CTNNB1 and prevents interaction between CTNNB1 and TCF family members. The encoded protein is a negative regulator of the Wnt signaling pathway. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ICAT
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9NSA3
CTNNBIP1
E. coli-derived recombinant human ICAT, Met1-Gln81.
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin)
536017
Unconjugated
CTNNBIP1
1110008O09Rik; 2310001I19Rik; AW457332; beta-catenin-interacting protein 1; beta-catenin-interacting protein ICAT; catenin; catenin beta interacting protein 1; catenin, beta interacting protein 1; catenin, beta-interacting protein 1; Catnbip1; Ctnnbip1; ICAT; inhibitor of beta-catenin and Tcf-4
Mouse
Protein A/G
RUO
56998
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.