missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
486.00€ - 679.00€
Specifications
| Antigen | ICB1 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18340235
|
Novus Biologicals
NBP3-17264-25UL |
25 μg |
486.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18358405
|
Novus Biologicals
NBP3-17264-100UL |
100 μg |
679.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICB1 Polyclonal antibody specifically detects ICB1 in Human samples. It is validated for ImmunofluorescenceSpecifications
| ICB1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| basement membrane-induced, chromosome 1 open reading frame 38, ICB1, ICB-1, Induced by contact to basement membrane 1 protein, Protein ICB-1, protein THEMIS2, THEMIS2, Thymocyte-expressed molecule involved in selection protein 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GEREENPEFTSLAVGDRLEVLGPGQAHGAQGSDVDVLVCQRLSDQAGEDEEEECKEEAESPERVLLPFHFPGSF | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 9473 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title