missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICOS (CD278) Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148519
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
Specifications
| ICOS (CD278) | |
| Polyclonal | |
| Unconjugated | |
| ICOS | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 54167 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Inhibition Assays, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9WVS0 | |
| ICOS | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ICOS, Met1-Leu142. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction