missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Tuning Solution I: Ba, Be, Ce, Co, In, Li, Mg, Pb, Rh, Tl, U, Y @ 10 ug/mL in 2% HNO3 / 5% HCl, VHG™

Product Code. 30003162
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003162 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30003162 Supplier VHG Labs Supplier No. VHGLPETSOL1100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-MS Tuning Solution I: Ba, Be, Ce, Co, In, Li, Mg, Pb, Rh, Tl, U, Y @ 10 ug/mL in 2% HNO3 / 5% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium/Beryllium/Cerium/Cobalt/Indium/Lead/Lithium/Magnesium/Rhodium/Thallium/Uranium/Yttrium
CAS 7440-39-3, 7440-41-7, 7440-45-1, 7440-48-4, 7440-74-6, 7439-92-1
Analyte or Component Names Barium/Beryllium/Cerium/Cobalt/Indium/Lead/Lithium/Magnesium/Rhodium/Thallium/Uranium/Yttrium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3/HCl
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Barium 1 μg/mL/Beryllium 1 μg/mL/Cerium 1 μg/mL/Cobalt 1 μg/mL/Indium 1 μg/mL/Lead 1 μg/mL/Lithium 1 μg/mL/Magnesium 1 μg/mL/Rhodium 1 μg/mL/Thallium 1 μg/mL/Uranium 1 μg/mL/Yttrium 1 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.