missing translation for 'onlineSavingsMsg'
Learn More

ICP-OES Low UV Wave Cal Solution: Al, P, S @ 10 μg/mL in 2% HNO3, VHG™

Product Code. 17924056
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924056 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17924056 Supplier VHG Labs Supplier No. VHGISUPELOW250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-OES Low UV Wave Cal Solution: Al, P, S @ 10 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Phosphorus/Sulfur
CAS 7429-90-5, 7723-14-0, 7704-34-9
Analyte or Component Names Aluminium/Phosphorus/Sulfur
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 250 mL
Concentration or Composition (by Analyte or Components) Aluminium 10 μg/mL/Phosphorus 10 μg/mL/Sulfur 10 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.