missing translation for 'onlineSavingsMsg'
Learn More

ICP-OES Multi-Element Setup Standard: As, K @ 50μg/mL; La, Li, Mn, Ni, Sr, Zn @ 10 μg/mL; Ba, Mg @ 1 μg/mL in 2% HNO3, VHG™

Product Code. 17932016
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932016 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932016 Supplier VHG Labs Supplier No. VHGISUPEOPTME500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-OES Multi-Element Setup Standard: As, K @ 50μg/mL; La, Li, Mn, Ni, Sr, Zn @ 10 μg/mL; Ba, Mg @ 1 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium/Magnesium/Lanthanum/Lithium/Manganese/Nickel/Strontium/Zirconium/Arsenic/Potassium
CAS 7440-39-3, 7439-95-4, 7439-91-0, 7439-93-2, 7439-96-5, 7440-02-0
Analyte or Component Names Barium/Magnesium/Lanthanum/Lithium/Manganese/Nickel/Strontium/Zirconium/Arsenic/Potassium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Barium 1 μg/mL/Magnesium 1 μg/mL/Lanthanum 10 μg/mL/Lithium 10 μg/mL/Manganese 10 μg/mL/Nickel 10 μg/mL/Strontium 10 μg/mL/Zirconium 10 μg/mL/Arsenic 50 μg/mL/Potassium 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.