missing translation for 'onlineSavingsMsg'
Learn More

IDH3B Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

129.00€ - 349.00€

Specifications

Antigen IDH3B
Clone 2D11A9
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234857
View Documents
Proteintech
68199-1-IG-20UL
20 μL
ONLINE EXCLUSIVE PRICE
145.00€
129.00€ / 20µL
Save 16.00€
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236284
View Documents
Proteintech
68199-1-IG-150UL
150 μL
ONLINE EXCLUSIVE PRICE
410.00€
349.00€ / 150µL
Save 61.00€
14% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The IDH3B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IDH3B. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The IDH3B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

IDH3B
1000 μg/mL
Monoclonal
Liquid
RUO
3420
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
2D11A9
Western Blot, ELISA
Unconjugated
Mouse
Human, Mouse, Rat, Rabbit, Pig
FLJ11043; H-IDHB; MGC903; RP46
IgG1
Protein G purification
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.