missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO Monoclonal Antibody (eyedio), APC-eFluor™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 47-9477-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
IDO Monoclonal antibody specifically detects IDO in Human samples. It is validated for Flow Cytometry
Specifications
| IDO | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| P14902 | |
| Ido1 | |
| Affinity chromatography | |
| RUO | |
| 3620 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| eyedio | |
| APC-eFluor 780 | |
| Ido1 | |
| EC 1.13.11.52; Ido; Ido1; IDO-1; Indo; indoleamine; Indoleamine 2,3-dioxygenase 1; Indoleamine-2; Indoleamine-pyrrole 2,3-dioxygenase | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction