Learn More
IDO2 Monoclonal Antibody (OTI2B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506646
Description
Along with the enzymes encoded by the INDO and TDO2 genes, the enzyme encoded by the INDOL1 gene metabolizes tryptophan in the kynurenine pathway.Specifications
| IDO2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZQW0 | |
| IDO2 | |
| Full length human recombit protein of human IDO2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2B9 | |
| Unconjugated | |
| IDO2 | |
| INDOL1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 169355 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.