missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFFO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16041-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody works well in WB, IHC and IF applications.
This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| IFFO1 | |
| Polyclonal | |
| Unconjugated | |
| IFFO1 | |
| HOM TES 103, IFFO, IFFO1, Tumor antigen HOM TES 103 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25900, 320678, 362437 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q0D2I5, Q8BXL9 | |
| Iffo1 | |
| IFFO1 Fusion Protein Ag8915 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction