missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12604-1-AP-20UL
This item is not returnable.
View return policy
Description
IFIT2 belongs to the IFIT family. It contains 6 TPR repeats. IFIT genes comprise a large family with three (Ifit1, Ifit2, and Ifit3) and four (IFIT1, IFIT2, IFIT3, and IFIT5) members in mice and humans, respectively. IFITproteins are produced in human body that are supposed to confer immunity against viral infection. These proteins are generally produced during viral infection, Interferon (IFN) treatment, and during pathogen recognition (Pathogen associated molecular pattern recognition) byimmune systemduring infections.Specifications
| IFIT2 | |
| Polyclonal | |
| Unconjugated | |
| IFIT2 | |
| cig42, G10P2, GARG 39, IFI 54, IFI 54K, IFI54, IFIT 2, IFIT2, ISG 54 K, ISG 54K, ISG54 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15958, 3433 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P09913, Q64112 | |
| IFIT2 | |
| IFIT2 Fusion Protein Ag3283 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction