missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha Antibody (967370), R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB93451
This item is not returnable.
View return policy
Description
IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for Block/Neutralize.
Specifications
| IFN-alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| IFNA2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IFN-alpha beta R1 Lys28-Lys436 Accession # AAA52730 | |
| 100 μg | |
| Cytokine Research | |
| 3440 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Blocking Assay | |
| 967370 | |
| Blocking/Neutralizing | |
| IFNA, IFNA2, IFNA2b, IFN-alphaA, INFA2, interferon alpha 2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction