missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma Antibody (154007) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB985M
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Porcine samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IFN-gamma | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Pig | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 154007 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| E. coli-derived recombinant porcine IFN-gamma , Ser21-Lys166, Accession # P17803 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
| 3458 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction