missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFNA17 Monoclonal Antibody (995826)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560844
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| IFNA17 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P01571 | |
| IFNA17 | |
| Human embryonic kidney cell line HEK293-derived recombinant human IFN-alpha I, Met1-Asp189. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Neutralization | |
| 995826 | |
| Unconjugated | |
| IFNA17 | |
| IFNA; IFNA17; IFN-alpha-17; IFN-alphaI; INFA; Interferon; interferon alpha 17; interferon alpha-17; Interferon alpha-88; interferon alpha-I'; Interferon alpha-T; interferon alpha-WA; interferon, alpha 17; leIF I; LEIF2C1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction