missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT80 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | IFT80 |
|---|---|
| Concentration | 0.44 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16823805
|
Proteintech
25230-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16813805
|
Proteintech
25230-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IFT80 gene encodes a protein with 7 WD40 domains that is a component of the intraflagellar transport. The IFT is essential for the development and maintece of motile and sensory cilia.Specifications
| IFT80 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8K057, Q9P2H3 | |
| 57560, 68259 | |
| IFT80 Fusion Protein Ag18650 | |
| Primary | |
| -20°C | |
| IFT80 |
| 0.44 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ATD2, IFT80, KIAA1374, WDR56 | |
| IFT80 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title