Learn More
Invitrogen™ IgD Monoclonal Antibody (IA6-2), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ H026T03R06-A
 View more versions of this product
Product Code. 30055375
  • 392.00€ / Pack of 100
Description
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgD
Monoclonal
0.2 μg/Test
PBS with 0.09% sodium azide; pH 7.2
P01880
IGHD
Affinity chromatography
RUO
3495
4°C, store in dark, DO NOT FREEZE!
Liquid
IgG2a κ
Flow Cytometry
IA6-2
NovaFluor Red 755
IGHD
IGHD; immunoglobulin heavy constant delta
Mouse
100 Tests