Learn More
Invitrogen™ Human IgD Monoclonal Antibody (IA6-2), NovaFluor™ UV 765, eBioscience™
Mouse Monoclonal Antibody
Brand: Invitrogen™ H026T03U01-A
Description
Description: This IA6-2 monoclonal antibody reacts with the heavy chain of human immunoglobulin D (IgD). This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding. This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer. Applications Tested: This IA6-2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test Master mixes.
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Specifications
| Human IgD | |
| Monoclonal | |
| 0.1 μg/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P01880 | |
| IGHD | |
| Affinity chromatography | |
| RUO | |
| 3495 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| IA6-2 | |
| NovaFluor UV 765 | |
| IGHD | |
| IGHD; immunoglobulin heavy constant delta | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.