Learn More
Invitrogen™ Human IgD Monoclonal Antibody (IA6-2), NovaFluor™ UV 765, eBioscience™
Mouse Monoclonal Antibody
176.00€ - 388.00€
Specifications
| Antigen | Human IgD |
|---|---|
| Clone | IA6-2 |
| Concentration | 0.1 μg/Test |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30274925
|
Invitrogen™
H026T02U01-A |
25 Tests |
176.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276109
|
Invitrogen™
H026T03U01-A |
100 Tests |
388.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This IA6-2 monoclonal antibody reacts with the heavy chain of human immunoglobulin D (IgD). This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding. This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer. Applications Tested: This IA6-2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test Master mixes.
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Specifications
| Human IgD | |
| 0.1 μg/Test | |
| Flow Cytometry | |
| NovaFluor UV 765 | |
| Mouse | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P01880 | |
| 3495 | |
| Primary | |
| Affinity chromatography |
| IA6-2 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Human | |
| IGHD | |
| IGHD; immunoglobulin heavy constant delta | |
| IGHD | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.