Learn More
IgD Monoclonal Antibody (IA6-2), Super Bright™ 780, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
156.00€ - 380.00€
Specifications
| Antigen | IgD |
|---|---|
| Clone | IA6-2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Super Bright 780 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16341944
|
eBioscience
78-9868-41 |
25 Tests | |||||||
16351944
![]() |
Invitrogen™
78-9868-42 |
100 Tests |
380.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The ISA-3 monoclonal antibody reacts with human ICOS (Inducible COStimulatory molecule), also known as H4, CRP-1 and AILIM. ICOS is a T cell specific activation molecule and a third member of the CD28/CTLA-4 family. Human ICOS has a relative molecular mass of 55-60 kDa, composed of 27 kDa and 29 kDa chains. Human ICOS on activated T cells has potent costimulatory activity for T cell activation and is required for humoral immune responses, in particular for memory B cell and plasma cell generation. ICOS binds to its ligand, B7h/B7RP-1 expressed on activated APCs (antigen presenting cells) and on a number of inflamed peripheral tissues. Plate-bound ISA-3 is costimulatory for T cells and induces production of IL-4, IL-5, IL-10 and other cytokines, but not IL-2. ISA-3 has the same reactivity pattern and characteristics as F44. ISA-3 was generated against the human ICOS antigen. C398.4A, anti-mouse ICOS/H4 (catalog No. 14-9949), was shown to cross-react with human ICOS but binds to an epitope different from ISA-3. C398.4A stains activated cells brighter than ISA-3; however, it also exhibits higher staining of non-activated human peripheral blood or isolated PBMC. To achieve the brightest staining of ICOS on activated human T cells, please use 13-9948 or 12-9948 rather than 11-9948. Applications Reported: This ISA-3 antibody has been reported for use in flow cytometric analysis.
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Specifications
| IgD | |
| Flow Cytometry | |
| Super Bright 780 | |
| Mouse | |
| Human | |
| 3495 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| IA6-2 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P01880 | |
| IGHD | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IGHD |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.