missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-I R/IGF1R Antibody (1H7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
216.00€ - 523.00€
Specifications
| Antigen | IGF-I R |
|---|---|
| Clone | 1H7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:40 |
| Applications | Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18267884
|
Novus Biologicals
NB100-66537-0.025MG |
0.025 mg |
216.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18202440
|
Novus Biologicals
NB100-66537 |
0.1 mg |
523.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGF-I R/IGF1R Monoclonal specifically detects IGF-I R/IGF1R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IGF-I R | |
| 1.0 mg/mL | |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer | |
| P08069 | |
| 3480 | |
| Purified human placental IGF-I receptor | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1H7 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:40 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF-I receptor, IGFIR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
| IGF1R | |
| IgG1 | |
| Protein G purified | |
| NB100-66537 recognizes human CD221 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title