missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF-II/IGF2 Antibody (8H1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-37462
This item is not returnable.
View return policy
Description
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IGF-II | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, PP9974, somatomedin-A | |
| Mouse | |
| 20.1 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication, Cytokine Research | |
| 3481 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8H1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| IGF2 | |
| Purified recombinant fragment of human IGF2 (AA: 25-180) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction