missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C1qA Antibody (C1QA/2783), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79915AF700
This item is not returnable.
View return policy
Description
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Complement Component C1qA | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C1QA | |
| Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) (Uniprot: P02745) | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| Human | |
| IgG2b κ |
| Western Blot, ELISA | |
| C1QA/2783 | |
| Western Blot, ELISA | |
| C1QA complement component 1, q subcomponent, A chain | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction