missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-2 Antibody (CL13563) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24568JF669
This item is not returnable.
View return policy
Description
IGFBP-2 Monoclonal antibody specifically detects IGFBP-2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| IGFBP-2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2 | |
| This antibody was developed using a synthetic peptide derived from P18065, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytokines and Growth Factors, Signal Transduction | |
| 3485 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL13563 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction