missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Antibody (rIG266), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79849H
This item is not returnable.
View return policy
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IgG | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| IGHG1 | |
| Recombinant full-length human IgG protein | |
| 0.1 mL | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with alpha (IgA), mu (IgM), epsilon (IgE), or delta (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This monoclonal antibody is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| rIG266 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Immunoglobulin G, ImmunoglobulinG | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction