missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (H, L) Rabbit anti-Dog, Polyclonal , Abnova™
Specifications
Specifications
| Antigen | IgG (H, L) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-dog IgG recognizes dog IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Liquid |
| Host Species | Rabbit |
| Immunogen | Dog IgG whole molecule |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Quantity | 10 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?