Learn More
IGJ Monoclonal Antibody (OTI3F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504175S
Description
IGJ Monoclonal antibody specifically detects IGJ in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| IGJ | |
| Monoclonal | |
| 0.73 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01591 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3512 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F8 | |
| Unconjugated | |
| JCHAIN | |
| 9530090F24Rik; AI323815; IGCJ; Igj; IgJ chain; immunoglobulin J chain; immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides; immunoglobulin joining chain; immunoglobulin joining gene (not J region); J chain; Jch; JCHAIN; Joining chain of multimeric IgA and IgM | |
| Full length human recombinant protein of human IGJ produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.