missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1B, p Thr1265 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42827AF532
This item is not returnable.
View return policy
Description
MAP1B Polyclonal specifically detects MAP1B in Human, Mouse, Rat, Chicken, Fish, Monkey, Reptile, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| MAP1B | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| DKFZp686E1099, DKFZp686F1345, FLJ38954, FUTSCH, MAP-1B, MAP5, microtubule-associated protein 1B | |
| Rabbit | |
| Affinity Purified | |
| Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Cell Markers | |
| 4131.0 | |
| Human, Mouse, Rat, Chicken, Fish, Monkey, Reptile, Xenopus species | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MAP1B | |
| Phosphopeptide made to a C-terminal portion of rat MAP1B. [Swiss-ProtNo. P15205] | |
| 0.1 mL | |
| Primary | |
| Phosphorylated Thr1265 on MAP1B (Thr1261 in rat). | |
| Store at 4°C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido