missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKEPP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
213.00€ - 450.00€
Specifications
| Antigen | IKEPP |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry reported in scientific literature (PMID 24391715), Immunohistochemistry-Paraffin, ICC/IF (Negative) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284420
|
Novus Biologicals
NB400-150SS |
0.025 mL |
213.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159219
|
Novus Biologicals
NB400-150 |
0.1 mL |
450.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IKEPP Polyclonal specifically detects IKEPP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).Specifications
| IKEPP | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Signal Transduction | |
| Whole antisera with 0.05% Sodium Azide | |
| FLJ22756, IKEPPPDZK2, intestinal and kidney enriched PDZ protein, Intestinal and kidney-enriched PDZ protein, Na(+)/H(+) exchange regulatory cofactor NHE-RF4, Na/Pi cotransporter C-terminal-associated protein 2, naPi-Cap2, Natrium-phosphate cotransporter IIa C-terminal-associated protein 2, NHERF4, NHERF-4, PDZ domain containing 2, PDZ domain containing 3, PDZ domain-containing protein 2, PDZ domain-containing protein 3, Sodium-hydrogen exchanger regulatory factor 4 | |
| PDZD3 | |
| IgG | |
| Unpurified | |
| 60 kDa |
| Western Blot 1:1000, Immunohistochemistry reported in scientific literature (PMID 24391715), Immunohistochemistry-Paraffin, ICC/IF (Negative) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| Q99MJ6 | |
| 79849 | |
| A synthetic peptide made to the C-terminus of mouse IKEPP. [UniProt# Q99MJ6] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title