missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Antibody (JES3-12G8) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
227.00€ - 457.00€
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | JES3-12G8 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot, ELISA 1:2000-1:4000, Immunohistochemistry 1:10 - 1:2000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10 - 1:2000, Enzyme-linked Immunosorbent Spot, Neutralization, ELISA Detection (Matched Antibody Pair) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISpot, Neutralization, ELISA Detection (Matched Antibody Pair) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18481242
|
Novus Biologicals
NBP2-27577 |
0.5 mg |
457.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18497651
|
Novus Biologicals
NBP2-27577-0.1mg |
0.1 mg |
227.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human, Cynomolgus Monkey, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Enzyme-linked Immunosorbent Spot, Neutralization, ELISA (Matched Antibody Pair)Specifications
| IL-10 | |
| 0.5 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ELISpot, Neutralization, ELISA Detection (Matched Antibody Pair) | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 1.0 mL of borate buffered saline, pH 8.2. | |
| CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| COS-expressed human IL-10 | |
| Primary | |
| Store at 4C. Do not freeze. |
| JES3-12G8 | |
| Western Blot, ELISA 1:2000-1:4000, Immunohistochemistry 1:10 - 1:2000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10 - 1:2000, Enzyme-linked Immunosorbent Spot, Neutralization, ELISA Detection (Matched Antibody Pair) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Cynomolgus Monkey, Virus | |
| NP_000563 | |
| 3586 | |
| IgG2a κ | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title