Learn More
IL-10R alpha Antibody (CL8203) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21164DL594
Product Code. 30307547
  • 1175.00€ / 0.10mL
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-10R alpha
Monoclonal
DyLight 594
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
This antibody was developed using a synthetic peptide derived from Q13651, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cytokine Research
3587
Store at 4⪚C in the dark.
IgG1
Western Blot
CL8203
50mM Sodium Borate
Mouse
Protein A purified