Learn More
IL-12 p70 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30756-1-AP-20UL
Description
The IL-12 p70 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse IL-12 p70. This antibody recognizes mouse antigen. The IL-12 p70 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-12 p70 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| IL12, IL-12, IL-12 p70 | |
| 20 μL | |
| 16160 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |