Learn More
Novus Biologicals™ IL-13 Antibody (CNTO 607), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28181-1mg
 View more versions of this product
Product Code. 30048288
  • 920.00€ / 1mg
Description
IL-13 Monoclonal antibody specifically detects IL-13 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-13
Monoclonal
Unconjugated
P35225
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
CNTO 607
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
IL-13
1 mg
Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
3596