missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-15 Antibody (34559), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2471J
This item is not returnable.
View return policy
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Western Blot, Neutralization, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| IL-15 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Neutralization, Flow Cytometry (Intracellular Staining), CyTOF | |
| 34559 | |
| Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| IL-15MGC9721, interleukin 15, interleukin-15 | |
| E. coli-derived recombinant human IL-15, Asn49-Ser162, Accession # P40933 | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3600 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction