Learn More
Novus Biologicals™ IL-17/IL-17A Antibody (CAT-2200), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28724-50ug
 View more versions of this product
Product Code. 30047242
  • 213.00€ / 50µg
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-17/IL-17A
Monoclonal
Unconjugated
Q16552
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
CAT-2200
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
CTLA-8/ IL-17a
50 μg
Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
3605