missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RA/IL-17R Antibody (CL10309) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21175DL594
This item is not returnable.
View return policy
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot
Specifications
| IL-17RA/IL-17R | |
| Monoclonal | |
| DyLight 594 | |
| CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| This antibody was developed using a synthetic peptide derived from Q96F46, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Innate Immunity | |
| 23765 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL10309 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction